CAS RN 39718-11-1
CAS RN 39718-11-1
IUPAC Name:
naphthalene-2,7-dicarbonitrile
Synonyms:
naphthalene-2,7-dicarbonitrile
Molecular Weight (g/mol):
178.19
Molecular Formula:
C12H6N2
InChi Key:
NNHXOKPHDUDDAK-UHFFFAOYSA-N
SMILES:
N#CC1=CC2=CC(=CC=C2C=C1)C#N
1
–
1
of
1
results
1
2,7-Dicyanonaphthalene 94.0+%, TCI America™
CAS: 39718-11-1 Molecular Formula: C12H6N2 Molecular Weight (g/mol): 178.194 InChI Key: NNHXOKPHDUDDAK-UHFFFAOYSA-N Synonym: 2,7-Naphthalenedicarbonitrile PubChem CID: 13301568 IUPAC Name: naphthalene-2,7-dicarbonitrile SMILES: C1=CC(=CC2=C1C=CC(=C2)C#N)C#N