accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOOH

CAS RN 3972-36-9

IUPAC Name: 2-benzylbutanedioic acid
Synonyms: benzylsuccinic acid β-carboxybenzenebutanoic acid
Molecular Weight (g/mol): 208.21
Molecular Formula: C11H12O4
InChi Key: GTOFKXZQQDSVFH-UHFFFAOYNA-N
SMILES: OC(=O)CC(CC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results