accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3973-70-4

IUPAC Name: 2-(4-aminopiperazin-1-yl)ethan-1-ol
Synonyms: 2-(4-aminopiperazin-1-yl)ethanol
Molecular Weight (g/mol): 145.21
Molecular Formula: C6H15N3O
InChi Key: XAOHWSFGKCJOGT-UHFFFAOYSA-N
SMILES: NN1CCN(CCO)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results