accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39741-46-3

IUPAC Name: 3-bromo-5-(chloromethyl)pyridine hydrochloride
Synonyms: 3-bromo-5-(chloromethyl)pyridine hydrochloride
Molecular Weight (g/mol): 242.93
Molecular Formula: C6H6BrCl2N
InChi Key: CJAUTVHMRSAZGG-UHFFFAOYSA-N
SMILES: Cl.ClCC1=CC(Br)=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results