accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39741-91-8

ICH3NCH3H3CI

CAS RN 39741-91-8

IUPAC Name: (iodomethyl)trimethylazanium iodide
Synonyms: (iodomethyl)trimethylazanium iodide
Molecular Weight (g/mol): 326.95
Molecular Formula: C4H11I2N
InChi Key: IFVIWWQMNYZYFQ-UHFFFAOYSA-M
SMILES: [I-].C[N+](C)(C)CI

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results