accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39745-40-9

IUPAC Name: 2-chloro-6-methylpyridin-3-amine
Synonyms: 2-chloro-6-methylpyridin-3-amine
Molecular Weight (g/mol): 142.59
Molecular Formula: C6H7ClN2
InChi Key: SUMSBUDMYMYTLL-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C(Cl)=N1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results