accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClClClClClCl

CAS RN 39765-80-5

IUPAC Name: 1,3,4,5,7,8,9,10,10-nonachlorotricyclo[5.2.1.0²,⁶]dec-8-ene
Synonyms: trans-nonachlor cis-nonachlor
Molecular Weight (g/mol): 444.2
Molecular Formula: C10H5Cl9
InChi Key: OCHOKXCPKDPNQU-UHFFFAOYNA-N
SMILES: ClC1C(Cl)C2C(C1Cl)C1(Cl)C(Cl)=C(Cl)C2(Cl)C1(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results