accessibility menu, dialog, popup

UserName

Viewing profile as user:
FNF(E/Z)

CAS RN 39769-09-0

IUPAC Name: N,1-bis(4-fluorophenyl)methanimine
Synonyms: N,1-bis(4-fluorophenyl)methanimine
Molecular Weight (g/mol): 217.22
Molecular Formula: C13H9F2N
InChi Key: FRNJXANITCYEMD-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=NC2=CC=C(F)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results