accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 397843-61-7

IUPAC Name: [4-(phenoxymethyl)phenyl]boronic acid
Synonyms: 4-(phenoxymethyl)phenylboronic acid
Molecular Weight (g/mol): 228.05
Molecular Formula: C13H13BO3
InChi Key: IXDHTLPOOZIZJX-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(COC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results