accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 397845-03-3

IUPAC Name: 4-bromo-2-methylbenzene-1-carbothioamide
Synonyms: 4-bromo-2-methylbenzenecarbothioamide
Molecular Weight (g/mol): 230.12
Molecular Formula: C8H8BrNS
InChi Key: KANLQQZRNHDXHE-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(Br)=C1)C(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results