accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3981-79-1

IUPAC Name: tetradecan-7-ol
Synonyms: tetradecan-7-ol
Molecular Weight (g/mol): 214.39
Molecular Formula: C14H30O
InChi Key: FFAQYGTZIMVBFJ-UHFFFAOYNA-N
SMILES: CCCCCCCC(O)CCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results