accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COH2N

CAS RN 39811-17-1

IUPAC Name: 4-methoxy-[1,1'-biphenyl]-3-amine
Synonyms: 4-methoxy-[1,1'-biphenyl]-3-amine
Molecular Weight (g/mol): 199.25
Molecular Formula: C13H13NO
InChi Key: DTYBRSLINXBXMP-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1N)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results