accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3982-91-0

IUPAC Name: phosphorothioyl trichloride
Synonyms: thiophosphoryl chloride
Molecular Weight (g/mol): 169.38
Molecular Formula: Cl3PS
InChi Key: WQYSXVGEZYESBR-UHFFFAOYSA-N
SMILES: ClP(Cl)(Cl)=S
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)