accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOCl

CAS RN 3984-34-7

IUPAC Name: 4-(4-chlorophenyl)-4-oxobutanoic acid
Synonyms: 4-(4-chlorophenyl)-4-oxobutanoic acid
Molecular Weight (g/mol): 212.63
Molecular Formula: C10H9ClO3
InChi Key: AHVASTJJVAYFPY-UHFFFAOYSA-N
SMILES: OC(=O)CCC(=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results