accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NBr

CAS RN 39856-58-1

IUPAC Name: 2-bromopyridin-3-amine
Synonyms: 2-bromopyridin-3-amine
Molecular Weight (g/mol): 173.01
Molecular Formula: C5H5BrN2
InChi Key: HKDVVTLISGIPFE-UHFFFAOYSA-N
SMILES: NC1=CC=CN=C1Br
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (4)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (3)