accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39878-87-0

IUPAC Name: hydrogen (2R)-2-amino-2-phenylacetyl chloride chloride
Synonyms: hydrogen (2R)-2-amino-2-phenylacetyl chloride chloride
Molecular Weight (g/mol): 206.07
Molecular Formula: C8H9Cl2NO
InChi Key: GVVFCAFBYHYGEE-OGFXRTJISA-N
SMILES: [H+].[Cl-].N[C@@H](C(Cl)=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)