accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39890-95-4

IUPAC Name: 2-chloro-6-(trifluoromethyl)pyridine
Synonyms: 2-chloro-6-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 181.54
Molecular Formula: C6H3ClF3N
InChi Key: ADVQMCQMDHBTHJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(Cl)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results