accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39891-12-8

IUPAC Name: 2-(5-bromopyridin-3-yl)acetic acid
Synonyms: (5-bromopyridin-3-yl)acetic acid
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: UETIDNDXXGCJCE-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(Br)=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results