accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOClN

CAS RN 39891-13-9

IUPAC Name: 2-(6-chloropyridin-3-yl)acetic acid
Synonyms: (6-chloropyridin-3-yl)acetic acid
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: XWVPSJPQWOVRHJ-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(Cl)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results