accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39896-06-5

IUPAC Name: 1-azabicyclo[2.2.2]octane hydrochloridyl
Synonyms: quinuclidine hydrochloridyl
Molecular Weight (g/mol): 146.64
Molecular Formula: C7H13ClN
InChi Key: ZXQNFZNRAGFIQH-UHFFFAOYSA-N
SMILES: [Cl].C1CN2CCC1CC2
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)