accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 399-35-9

IUPAC Name: N-(3-chloro-4-fluorophenyl)acetamide
Synonyms: N-(3-chloro-4-fluorophenyl)acetamide
Molecular Weight (g/mol): 187.6
Molecular Formula: C8H7ClFNO
InChi Key: ALPHMTFVUKDBGJ-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(F)C(Cl)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results