accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 399-51-9

IUPAC Name: 6-fluoro-1H-indole
Synonyms: 6-fluoro-1H-indole
Molecular Weight (g/mol): 135.14
Molecular Formula: C8H6FN
InChi Key: YYFFEPUCAKVRJX-UHFFFAOYSA-N
SMILES: FC1=CC=C2C=CNC2=C1
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (5)
  • (3)
  • (1)
  • (1)
  • (2)
  • (5)