accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 399-74-6

IUPAC Name: 2-chloro-6-fluoro-1,3-benzothiazole
Synonyms: 2-chloro-6-fluoro-1,3-benzothiazole
Molecular Weight (g/mol): 187.62
Molecular Formula: C7H3ClFNS
InChi Key: ISIIQFDYFMPPOA-UHFFFAOYSA-N
SMILES: FC1=CC=C2N=C(Cl)SC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results