accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NF

CAS RN 399-88-2

IUPAC Name: 3-fluoro-4-methylpyridine
Synonyms: 3-fluoro-4-methylpyridine
Molecular Weight (g/mol): 111.12
Molecular Formula: C6H6FN
InChi Key: WZMOEPZZTTWDIA-UHFFFAOYSA-N
SMILES: CC1=C(F)C=NC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results