accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONHOSCH3HCl

CAS RN 399017-10-8

IUPAC Name: ethyl 4-methyl-2-(4-oxocyclohexa-2,5-dien-1-ylidene)-2,3-dihydro-1,3-thiazole-5-carboxylate hydrochloride
Synonyms: ethyl 4-methyl-2-(4-oxocyclohexa-2,5-dien-1-ylidene)-3H-1,3-thiazole-5-carboxylate hydrochloride
Molecular Weight (g/mol): 299.77
Molecular Formula: C13H14ClNO3S
InChi Key: CTNLNGHJGMEXNQ-UHFFFAOYSA-N
SMILES: Cl.CCOC(=O)C1=C(C)NC(S1)=C1C=CC(=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results