accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONH2

CAS RN 39905-57-2

IUPAC Name: 4-(hexyloxy)aniline
Synonyms: 4-hexyloxyaniline
Molecular Weight (g/mol): 193.29
Molecular Formula: C12H19NO
InChi Key: DJRKHTCUXRGYEU-UHFFFAOYSA-N
SMILES: CCCCCCOC1=CC=C(N)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results