accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3

CAS RN 3992-48-1

IUPAC Name: 8-(acetyloxy)octyl acetate
Synonyms: 8-(acetyloxy)octyl acetate
Molecular Weight (g/mol): 230.3
Molecular Formula: C12H22O4
InChi Key: MDHIGESKEUQAEO-UHFFFAOYSA-N
SMILES: CC(=O)OCCCCCCCCOC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results