CAS RN 3997-89-5
IUPAC Name:
cyclopentanone
Synonyms:
cyclopentanone ketopentamethylene adipic ketone adipinketon dumasin
Molecular Weight (g/mol):
84.12
Molecular Formula:
C5H8O
InChi Key:
BGTOWKSIORTVQH-UHFFFAOYSA-N
SMILES:
O=C1CCCC1
1
–
1
of
1
results
1
Promotions Available
Cyclopentanone-2,2,5,5-d{4}, 95%
Cyclopentanone-2,2,5,5-d{4}, 95%
CAS: 3997-89-5 Molecular Formula: C5H8O Molecular Weight (g/mol): 88.142 MDL Number: MFCD00190415 InChI Key: BGTOWKSIORTVQH-KHORGVISSA-N Synonym: cyclopentanone-2,2,5,5-d4,cyclopentanone d4,2,2,5,5-∼2∼h_4_ cyclopentanone,cyclopentanone-2,2,5,5-d4, 98 atom % d,2,2,5,5-2 h? cyclopentan-1-one PubChem CID: 16213326 IUPAC Name: 2,2,5,5-tetradeuteriocyclopentan-1-one SMILES: C1CCC(=O)C1