accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3999-38-0

IUPAC Name: 2-methylpyridine borane
Synonyms: α-methylpyridine borane
Molecular Weight (g/mol): 106.96
Molecular Formula: C6H10BN
InChi Key: ZMASBEFBIXMNCP-UHFFFAOYSA-N
SMILES: B.CC1=CC=CC=N1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)