accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3H3COOFFF

CAS RN 400-53-3

IUPAC Name: trimethylsilyl 2,2,2-trifluoroacetate
Synonyms: trimethylsilyl 2,2,2-trifluoroacetate
Molecular Weight (g/mol): 186.21
Molecular Formula: C5H9F3O2Si
InChi Key: VIYXXANHGYSBLY-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OC(=O)C(F)(F)F
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)