accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOClClFFF

CAS RN 400-70-4

IUPAC Name: 1,5-dichloro-2-nitro-4-(trifluoromethyl)benzene
Synonyms: 1,5-dichloro-2-nitro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 259.99
Molecular Formula: C7H2Cl2F3NO2
InChi Key: VLVNHMVSVDVAOA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=C(Cl)C=C1Cl)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results