accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COFNOO

CAS RN 400-93-1

IUPAC Name: 1-(4-fluoro-3-nitrophenyl)ethan-1-one
Synonyms: 1-(4-fluoro-3-nitrophenyl)ethanone
Molecular Weight (g/mol): 183.14
Molecular Formula: C8H6FNO3
InChi Key: PTCNZDJJIOLIKQ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(F)C(=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)