accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNOOFFF

CAS RN 400-99-7

IUPAC Name: 2-nitro-4-(trifluoromethyl)phenol
Synonyms: 2-nitro-4-(trifluoromethyl)phenol
Molecular Weight (g/mol): 207.11
Molecular Formula: C7H4F3NO3
InChi Key: XZEDEVRSUANQEM-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1[N+]([O-])=O)C(F)(F)F
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)