accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SNBrOOH

CAS RN 40003-41-6

IUPAC Name: 2-bromo-4-methyl-1,3-thiazole-5-carboxylic acid
Synonyms: 2-bromo-4-methyl-1,3-thiazole-5-carboxylic acid
Molecular Weight (g/mol): 222.06
Molecular Formula: C5H4BrNO2S
InChi Key: HMSQZHBSTZZNGI-UHFFFAOYSA-N
SMILES: CC1=C(SC(Br)=N1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results