accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONNH

CAS RN 40004-08-8

IUPAC Name: ethyl 2-(piperazin-1-yl)acetate
Synonyms: ethyl 2-(piperazin-1-yl)acetate
Molecular Weight (g/mol): 172.23
Molecular Formula: C8H16N2O2
InChi Key: MTFCXMJOGMHYAE-UHFFFAOYSA-N
SMILES: CCOC(=O)CN1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results