accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SNOHO

CAS RN 40004-69-1

IUPAC Name: 2-methyl-1,3-thiazole-5-carboxylic acid
Synonyms: 2-methyl-1,3-thiazole-5-carboxylic acid
Molecular Weight (g/mol): 143.16
Molecular Formula: C5H5NO2S
InChi Key: QCXCIYPOMMIBHO-UHFFFAOYSA-N
SMILES: CC1=NC=C(S1)C(O)=O
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results