accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 400071-95-6

IUPAC Name: 5-bromo-1-methyl-1H-indole-3-carboxylic acid
Synonyms: 5-bromo-1-methylindole-3-carboxylic acid
Molecular Weight (g/mol): 254.08
Molecular Formula: C10H8BrNO2
InChi Key: WQZLFFJMRKTCMU-UHFFFAOYSA-N
SMILES: CN1C=C(C(O)=O)C2=CC(Br)=CC=C12
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results