accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40020-87-9

IUPAC Name: 1-(7-hydroxy-1-benzofuran-2-yl)ethan-1-one
Synonyms: 1-(7-hydroxy-1-benzofuran-2-yl)ethanone
Molecular Weight (g/mol): 176.17
Molecular Formula: C10H8O3
InChi Key: HRIVIHCUSKDPNK-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC2=CC=CC(O)=C2O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results