accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSBr

CAS RN 40032-73-3

IUPAC Name: 3-bromo-2-chlorothiophene
Synonyms: 3-bromo-2-chlorothiophene
Molecular Weight (g/mol): 197.47
Molecular Formula: C4H2BrClS
InChi Key: KSHOQKKCPJELBV-UHFFFAOYSA-N
SMILES: ClC1=C(Br)C=CS1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results