accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4005-51-0

IUPAC Name: 1,3,4-thiadiazol-2-amine
Synonyms: 2-amino-1,3,4-thiadiazole
Molecular Weight (g/mol): 101.13
Molecular Formula: C2H3N3S
InChi Key: QUKGLNCXGVWCJX-UHFFFAOYSA-N
SMILES: NC1=NN=CS1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)