accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40052-13-9

IUPAC Name: 3-(tert-butoxy)-3-oxopropanoic acid
Synonyms: 3-(tert-butoxy)-3-oxopropanoic acid
Molecular Weight (g/mol): 160.17
Molecular Formula: C7H12O4
InChi Key: NGGGZUAEOKRHMA-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)CC(O)=O
Boiling Point 
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)