accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrOH

CAS RN 40054-01-1

IUPAC Name: [6-(bromomethyl)pyridin-2-yl]methanol
Synonyms: [6-(bromomethyl)pyridin-2-yl]methanol
Molecular Weight (g/mol): 202.05
Molecular Formula: C7H8BrNO
InChi Key: JLYYQZROGJLCCS-UHFFFAOYSA-N
SMILES: OCC1=NC(CBr)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results