accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNNOO

CAS RN 4008-48-4

IUPAC Name: 5-nitroquinolin-8-ol
Synonyms: nitroxoline nicene forte noxibiol 5-nitroxine 5-nok nitroxolina [inn-spanish] 5-nitrox nitroxoline (inn) and its salts nitroxolinum 5-nitroks
Molecular Weight (g/mol): 190.16
Molecular Formula: C9H6N2O3
InChi Key: RJIWZDNTCBHXAL-UHFFFAOYSA-N
SMILES: OC1=C2N=CC=CC2=C(C=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)