accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCH3OP

CAS RN 4009-98-7

IUPAC Name: (methoxymethyl)triphenylphosphanium chloride
Synonyms: (methoxymethyl)triphenylphosphanium chloride
Molecular Weight (g/mol): 342.8
Molecular Formula: C20H20ClOP
InChi Key: SJFNDMHZXCUXSA-UHFFFAOYSA-M
SMILES: [Cl-].COC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (9)
Quantity 
  • (4)
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Physical Form 
  • (6)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (3)
  • (6)
  • (2)
  • (9)
  • (4)
  • (4)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)