accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFOFFF

CAS RN 401-95-6

IUPAC Name: 3,5-bis(trifluoromethyl)benzaldehyde
Synonyms: 3,5-bis(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 242.12
Molecular Formula: C9H4F6O
InChi Key: LDWLIXZSDPXYDR-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(C=O)=C1)C(F)(F)F
Boiling Point 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)