accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOFFFNOO

CAS RN 401-99-0

IUPAC Name: 1,3-dinitro-5-(trifluoromethyl)benzene
Synonyms: 1,3-dinitro-5-(trifluoromethyl)benzene
Molecular Weight (g/mol): 236.11
Molecular Formula: C7H3F3N2O4
InChi Key: QZADIXWDDVQVKM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC(=C1)[N+]([O-])=O)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results