accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNONNH2NH2NH2NOOHClHCl

CAS RN 40127-11-5

IUPAC Name: 2-amino-5-[(diaminomethylidene)amino]-N-(4-nitrophenyl)pentanamide dihydrochloride
Synonyms: 2-amino-5-[(diaminomethylidene)amino]-N-(4-nitrophenyl)pentanamide dihydrochloride
Molecular Weight (g/mol): 367.23
Molecular Formula: C12H20Cl2N6O3
InChi Key: FBVMDLFQVOFFHS-UHFFFAOYNA-N
SMILES: Cl.Cl.NC(CCCN=C(N)N)C(=O)NC1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)