accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNH2OHO

CAS RN 40155-43-9

IUPAC Name: 5-aminopyrazine-2-carboxylic acid
Synonyms: 5-aminopyrazine-2-carboxylic acid
Molecular Weight (g/mol): 139.11
Molecular Formula: C5H5N3O2
InChi Key: FMEFOOOBIHQRMK-UHFFFAOYSA-N
SMILES: NC1=NC=C(N=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results