accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFBr

CAS RN 40161-55-5

IUPAC Name: 1-bromo-4-fluoro-2-(trifluoromethyl)benzene
Synonyms: 1-bromo-4-fluoro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 243
Molecular Formula: C7H3BrF4
InChi Key: AIDVAZGOACECLJ-UHFFFAOYSA-N
SMILES: FC1=CC(=C(Br)C=C1)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results