accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOHNH3C

CAS RN 401647-22-1

IUPAC Name: methyl[(3-methyl-1,2-oxazol-5-yl)methyl]amine
Synonyms: methyl[(3-methyl-1,2-oxazol-5-yl)methyl]amine
Molecular Weight (g/mol): 126.16
Molecular Formula: C6H10N2O
InChi Key: UQKWKRSUELRKSK-UHFFFAOYSA-N
SMILES: CNCC1=CC(C)=NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results